Anti-SARS-CoV-2 Spike Protein (RBD) Human Monoclonal Antibody [clone: HC2001]

Supplier: Genscript
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A02038-100 A02038-1
GENSA02038-100EA 586 GBP
GENSA02038-100 GENSA02038-1
Anti-SARS-CoV-2 Spike Protein (RBD) Human Monoclonal Antibody [clone: HC2001]
Antibodies
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.
SARS-CoV-2 Spike S1 Antibody (HC2001), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.

Type: Primary
Antigen: Protein
Clonality: Monoclonal
Clone: HC2001 (More Product Information
Order Now

SPECIFICATIONS

Antigen name SARS-CoV-2 Spike Protein (RBD)
Environmentally Preferable
Antigen synonyms Receptor-binding domain/RBD
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone HC2001
Reactivity Human
Host Human
Gene ID MN908947
Isotype IgG1
ELISA Yes
Antibody fragment Whole Molecule
Modification Humanization
Epitope Confidentiality
Immunogen Confidentiality
Amino acid number Confidentiality
Sequence Confidentiality
Format Purified antibody
Form liquid
Molecular weight 150 kDa
Purification Protein A
Concentration 1 mg/ml, supplied in PBS, pH 7.4, containing 0.02% sodium azide.
Cross adsorption No
Storage buffer PBS, pH 7.4, containing 0.02% sodium azide.
Storage temperature ­20° C
Shipping temperature 4° C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR