Anti-cGFP-tag Mouse Monoclonal Antibody

Supplier: Genscript
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A00185-40
GENSA00185-40EA 163 GBP
GENSA00185-40
Anti-cGFP-tag Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: cGFP-tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR