DNA ladder, 1 kb Plus, DNAmark™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-854
GENO786-854EA 215 GBP
GENO786-854
DNA ladder, 1 kb Plus, DNAmark™
Electrophoresis Reagents Molecular Weight Markers
DNAmark™ 1 kb Plus ladder contains 15 DNA bands ranging from 100 bp to 10 kb. The 500 bp and 3000 bp fragment are present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp and 3000 bp are 40 ng. The 500 bp and 3000 bp fragment are 100 ng.
  • Pre-mixed with loading dye and ready to use