Human Serum Albumin (HSA) Powder
Supplier: GEMINI BIO-PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
800-125P-001 800-125P-005 800-125P-025
GEMB800-125P-001EA 92 GBP
GEMB800-125P-001 GEMB800-125P-005 GEMB800-125P-025
Human Serum Albumin (HSA) Powder
Sera (Serum)
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilised. MW ~66 kDa.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.