DMSO resistant peel heat seal
Supplier: 4titude Ltd.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4TI-0586-DMSO 4TI-0585 4TI-0587
FTIT4TI-0586-DMSOEA 1320.75 GBP
FTIT4TI-0586-DMSO FTIT4TI-0585 FTIT4TI-0587
DMSO resistant peel heat seal
Seals Microplate Seals
Solvent resistant heat sealing foil suitable for low and room temperature compound storage.
  • Peelable from PP and COC plates, forms a weld seal to PE plates
  • Temperature range –80 to +40 °C