(4-Aminophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Aminophenylacetic acid

043462-100G
FLUO043462-100GEA 0 GBP
FLUO043462-100G
(4-Aminophenyl)acetic acid
(4-Aminophenyl)acetic acid
Formula: (C₆H₄(NH₂))CH₂COOH
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 199…202 °C
MDL Number: MFCD00007916
CAS Number: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR