3-Amino-2-fluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

033249-5G
FLUO033249-5GEA 0 GBP
FLUO033249-5G
3-Amino-2-fluoropyridine
3-Amino-2-fluoropyridine
Formula: C₅H₅FN₂
MDL Number: MFCD03095248
CAS Number: 1597-33-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR