2'-Isopropyl-2,2,2-trifluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,2,2-Trifluoro-1-(2-isopropylphenyl)ethanone

023163-1G
FLUO023163-1GEA 0 GBP
FLUO023163-1G
2'-Isopropyl-2,2,2-trifluoroacetophenone
2'-Isopropyl-2,2,2-trifluoroacetophenone
Formula: C₁₁H₁₁F₃O
MW: 216.2 g/mol
MDL Number: MFCD01319977
CAS Number: 845823-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR