4-Butyl-3',4'-dichlorobenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

022776-1G
FLUO022776-1GEA 0 GBP
FLUO022776-1G
4-Butyl-3',4'-dichlorobenzophenone
4-Butyl-3',4'-dichlorobenzophenone
Formula: C₁₇H₁₆Cl₂O
MDL Number: MFCD06201541
CAS Number: 844885-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR