4-Methoxythiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Methoxybenzothioamide

021331-1G 021331-25G 021331-5G
FLUO021331-1GEA 0 GBP
FLUO021331-1G FLUO021331-25G FLUO021331-5G
4-Methoxythiobenzamide
4-Methoxythiobenzamide
Formula: C₈H₉NOS
MW: 167.23 g/mol
Boiling Pt: 282
Melting Pt: 155
MDL Number: MFCD00040993
CAS Number: 2362-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR