2-Chlorothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

021317-1G 021317-2.5G 021317-5G
FLUO021317-1GEA 0 GBP
FLUO021317-1G FLUO021317-2.5G FLUO021317-5G
2-Chlorothiobenzamide
2-Chlorothiobenzamide
Formula: C₇H₆ClNS
MW: 171.65 g/mol
Boiling Pt: 316
Melting Pt: 109…110 °C
MDL Number: MFCD00040955
CAS Number: 15717-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR