4-Methoxy-m-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

018789-5G
FLUO018789-5GEA 0 GBP
FLUO018789-5G
4-Methoxy-m-toluic acid
4-Methoxy-m-toluic acid
Formula: C₉H₁₀O₃
MDL Number: MFCD00270114
CAS Number: 6880-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR