3,4-Dimethoxybenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

018572-10G
FLUO018572-10GEA 0 GBP
FLUO018572-10G
3,4-Dimethoxybenzoyl chloride
3,4-Dimethoxybenzoyl chloride
Formula: C₉H₉ClO₃
Boiling Pt: 95…98 °C (1 mmHg)
Melting Pt: 71 °C
MDL Number: MFCD00000674
CAS Number: 3535-37-3
EINECS: 222-568-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR