(4-Aminophenylthio)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(4-Aminophenylthio)acetic acid

018190-10G
FLUO018190-10GEA 0 GBP
FLUO018190-10G
(4-Aminophenylthio)acetic acid
(4-Aminophenylthio)acetic acid
Formula: C₈H₉NO₂S
MW: 183.23 g/mol
MDL Number: MFCD00025360
CAS Number: 104-18-7
EINECS: 203-182-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR