2-Phenylethylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Phenethylboronic acid

016267-1G 016267-5G
FLUO016267-1GEA 0 GBP
FLUO016267-1G FLUO016267-5G
2-Phenylethylboronic acid
2-Phenylethylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01631226
CAS Number: 34420-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR