2'-Fluorobiphenyl-4-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid

014264-100MG 014264-1G
FLUO014264-100MGEA 0 GBP
FLUO014264-100MG FLUO014264-1G
2'-Fluorobiphenyl-4-carboxylic acid
2'-Fluorobiphenyl-4-carboxylic acid
Formula: C₁₃H₉FO₂
MDL Number: MFCD00278732
CAS Number: 365-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR