4-Bromo-3,5-xylenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene

013945-100G 013945-25G
FLUO013945-100GEA 0 GBP
FLUO013945-100G FLUO013945-25G
4-Bromo-3,5-xylenol
4-Bromo-3,5-xylenol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 259
Melting Pt: 116
MDL Number: MFCD00002315
CAS Number: 7463-51-6
EINECS: 231-255-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR