3-Bromo-α,α,α-trifluoro-5-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

013787-1G
FLUO013787-1GEA 0 GBP
FLUO013787-1G
3-Bromo-α,α,α-trifluoro-5-nitrotoluene
3-Bromo-α,α,α-trifluoro-5-nitrotoluene
Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR