(±)-2-Fluoro-α-methylbenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-(2-Fluorophenyl)ethylamine

013377-1G
FLUO013377-1GEA 0 GBP
FLUO013377-1G
(±)-2-Fluoro-α-methylbenzylamine
(±)-2-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MDL Number: MFCD04038307
CAS Number: 74788-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR