3-Fluoro-2-methoxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

013029-1G 013029-5G
FLUO013029-1GEA 0 GBP
FLUO013029-1G FLUO013029-5G
3-Fluoro-2-methoxybenzoic acid
3-Fluoro-2-methoxybenzoic acid
Formula: C₈H₇FO₃
MDL Number: MFCD00973972
CAS Number: 106428-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR