6-Methoxy-1-indanone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

012751-1G 012751-25G 012751-5G
FLUO012751-1GEA 0 GBP
FLUO012751-1G FLUO012751-25G FLUO012751-5G
6-Methoxy-1-indanone
6-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 270
Melting Pt: 108
MDL Number: MFCD00021232
CAS Number: 13623-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR