5-Bromo-2,3-dichloropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

012745-1G 012745-5G
FLUO012745-1GEA 0 GBP
FLUO012745-1G FLUO012745-5G
5-Bromo-2,3-dichloropyridine
5-Bromo-2,3-dichloropyridine
Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
MDL Number: MFCD04112492
CAS Number: 97966-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR