2,6-Dichloronicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

011683-5G
FLUO011683-5GEA 0 GBP
FLUO011683-5G
2,6-Dichloronicotinic acid
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR