3-Formylfuran-2-boronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (3-Formylfuran-2-yl)boronic acid

011037-5G 011037-1G
FLUO011037-5GEA 0 GBP
FLUO011037-5G FLUO011037-1G
3-Formylfuran-2-boronic acid
3-Formylfuran-2-boronic acid
Formula: C₅H₅BO₄
MW: 139.90 g/mol
MDL Number: MFCD01075680
CAS Number: 27339-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR