5-Chloro-2-methoxyphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

011022-5G
FLUO011022-5GEA 0 GBP
FLUO011022-5G
5-Chloro-2-methoxyphenylboronic acid
5-Chloro-2-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MDL Number: MFCD01318966
CAS Number: 89694-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR