3-Chlorophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

010985-25G 010985-5G
FLUO010985-25GEA 0 GBP
FLUO010985-25G FLUO010985-5G
3-Chlorophenylboronic acid
3-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR