Sodium-1-decanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

009634-5G 009634-25G
FLUO009634-5GEA 0 GBP
FLUO009634-5G FLUO009634-25G
Sodium-1-decanesulphonate
Sodium 1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
MW: 244.33 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR