3-(2-Fluorophenyl)propionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

009235-5G 009235-25G 009235-1G
FLUO009235-5GEA 0 GBP
FLUO009235-5G FLUO009235-25G FLUO009235-1G
3-(2-Fluorophenyl)propionic acid
3-(2-Fluorophenyl)propionic acid
Formula: C₉H₉FO₂
MDL Number: MFCD01310820
CAS Number: 1643-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR