5-Bromo-2-fluorobenzylamine hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (5-Bromo-2-fluorophenyl)methanamine hydrochloride

008871-1G 008871-5G
FLUO008871-1GEA 0 GBP
FLUO008871-1G FLUO008871-5G
5-Bromo-2-fluorobenzylamine hydrochloride
5-Bromo-2-fluorobenzylamine hydrochloride
Formula: C₇H₈BrClFN
MW: 240.50 g/mol
MDL Number: MFCD00143426
CAS Number: 202865-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR