2,6-Diethylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1,3-Diethyl-2-isothiocyanatobenzene

008865-5G 008865-25G
FLUO008865-5GEA 0 GBP
FLUO008865-5G FLUO008865-25G
2,6-Diethylphenyl isothiocyanate
2,6-Diethylphenyl isothiocyanate
Formula: C₁₁H₁₃NS
MW: 191.3 g/mol
Boiling Pt: 82 °C (0.3 mmHg)
MDL Number: MFCD00041069
CAS Number: 25343-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR