2-Hydroxythiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: o-Hydroxythiophenol, 2-Hydroxyphenyl mercaptan, 2-Hydroxybenzene-1-thiol, 1-Hydroxy-2-mercaptobenzene, 2-Mercaptophenol, 2-Hydroxybenzenethiol

FL8733.1 008733-5G 008733-25G
CHMPFL8733.1EA 0 GBP
CHMPFL8733.1 FLUO008733-5G FLUO008733-25G
2-Hydroxythiophenol
2-Hydroxythiophenol
Formula: C₆H₆OS
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00040447
CAS Number: 1121-24-0
EINECS: 214-326-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR