5-Fluoro-2-(trifluoromethyl)cinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

007934-1G 007934-5G
FLUO007934-1GEA 0 GBP
FLUO007934-1G FLUO007934-5G
5-Fluoro-2-(trifluoromethyl)cinnamic acid
5-Fluoro-2-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236299
CAS Number: 231291-18-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR