3-Fluoro-4-(trifluoromethyl)benzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-(Bromomethyl)-3-fluoro-4-(trifluoromethyl)benzene, 4-(Bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene

007924-1G 007924-5G
FLUO007924-1GEA 0 GBP
FLUO007924-1G FLUO007924-5G
3-Fluoro-4-(trifluoromethyl)benzyl bromide
3-Fluoro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 197
Melting Pt: 22
MDL Number: MFCD00236289
CAS Number: 213203-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR