α,α,α,5-Tetrafluoro-o-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

007912-5G 007912-25G
FLUO007912-5GEA 0 GBP
FLUO007912-5G FLUO007912-25G
α,α,α,5-Tetrafluoro-o-toluic acid
α,α,α,5-Tetrafluoro-o-toluic acid
Formula: C₈H₄F₄O₂
MDL Number: MFCD00083521
CAS Number: 654-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR