3'-Fluoro-4'-(trifluoromethyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanone

007905-5G 007905-1G
FLUO007905-5GEA 0 GBP
FLUO007905-5G FLUO007905-1G
3'-Fluoro-4'-(trifluoromethyl)acetophenone
3'-Fluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MDL Number: MFCD00236273
CAS Number: 237761-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR