2,3,4-Trifluorobenzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene

007503-1G 007503-5G 007503-10G
FLUO007503-1GEA 0 GBP
FLUO007503-1G FLUO007503-5G FLUO007503-10G
2,3,4-Trifluorobenzyl bromide
2,3,4-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 182
Melting Pt: 21
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00061233
CAS Number: 157911-55-2
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR