1,1,1-Trifluoroacetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

007468-25G 007468-100G
FLUO007468-25GEA 0 GBP
FLUO007468-25G FLUO007468-100G
1,1,1-Trifluoroacetone
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
Boiling Pt: 22 °C (1013 hPa)
Melting Pt: –78 °C
Density: 1,18 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 1993
ADR: 3,I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR