Titanium(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Titanium trifluoride

007427-5G 007427-25G 007427-100G
FLUO007427-5GEA 0 GBP
FLUO007427-5G FLUO007427-25G FLUO007427-100G
Titanium(III) fluoride
Titanium (III) fluoride
Formula: F₃Ti
Density: 2.98 g/cm³ (20 °C)
MDL Number: MFCD00042544
CAS Number: 13470-08-1
EINECS: 236-732-4
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR