Sodium tetrafluoroborate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium fluoroborate

007291-25G 007291-500G 007291-10KG 007291-1KG
FLUO007291-25GEA 0 GBP
FLUO007291-25G FLUO007291-500G FLUO007291-10KG FLUO007291-1KG
Sodium tetrafluoroborate
Sodium tetrafluoroborate
Formula: BF₄Na
Melting Pt: 384 °C
Density: ∼2.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
Merck Index: 12,08761

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR