4,4,5,5,5-Pentafluoropentan-1-ol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4,4,5,5,5-Pentafluoropentanol

006963-5G 006963-25G 006963-100G
FLUO006963-5GEA 0 GBP
FLUO006963-5G FLUO006963-25G FLUO006963-100G
4,4,5,5,5-Pentafluoropentan-1-ol
4,4,5,5,5-Pentafluoropentan-1-ol
Formula: C₅H₇F₅O
MDL Number: MFCD00153224
CAS Number: 148043-73-6
EINECS: 421-360-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR