N-Methyl-2-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

006836-1G 006836-5G
FLUO006836-1GEA 0 GBP
FLUO006836-1G FLUO006836-5G
N-Methyl-2-(trifluoromethoxy)aniline
N-Methyl-2-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO
MDL Number: MFCD00052334
CAS Number: 175278-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR