Hafnium(IV) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

006500-25G
FLUO006500-25GEA 0 GBP
FLUO006500-25G
Hafnium(IV) fluoride
Hafnium (IV) fluoride
Formula: F₄Hf
MDL Number: MFCD00016127
CAS Number: 13709-52-9
EINECS: 237-258-0
UN: 3290
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR