5-Fluoro-o-toluonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

006279-1G 006279-5G
FLUO006279-1GEA 0 GBP
FLUO006279-1G FLUO006279-5G
5-Fluoro-o-toluonitrile
5-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 226
Melting Pt: 42…46 °C
MDL Number: MFCD00042295
CAS Number: 77532-79-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR