2-Fluoronicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

005467-1G 005467-5G 005467-25G
FLUO005467-1GEA 0 GBP
FLUO005467-1G FLUO005467-5G FLUO005467-25G
2-Fluoronicotinic acid
2-Fluoronicotinic acid
Formula: C₆H₄FNO₂
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR