3-Bromo-4-fluorocinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

004224-5G
FLUO004224-5GEA 0 GBP
FLUO004224-5G
3-Bromo-4-fluorocinnamic acid
3-Bromo-4-fluorocinnamic acid
Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 189…192 °C
MDL Number: MFCD00143095
CAS Number: 160434-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR