2,4-Difluorobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CFF611.5 003579-5G 003579-10G 003579-25G
CHMPCFF611.5EA 0 GBP
CHMPCFF611.5 FLUO003579-5G FLUO003579-10G FLUO003579-25G
2,4-Difluorobenzenesulphonyl chloride
2,4-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
Boiling Pt: 84 °C (1013 hPa)
Density: 1.581 g/cm³ (20 °C)
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR