2-Chlorobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CFF434.10 CFF434.50 CFF434.100 003573-5G 003573-25G
CHMPCFF434.10EA 0 GBP
CHMPCFF434.10 CHMPCFF434.50 CHMPCFF434.100 FLUO003573-5G FLUO003573-25G
2-Chlorobenzenesulphonyl chloride
2-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 88…92 °C (0.1 mmHg)
Melting Pt: 28 °C
Density: 1.548 g/cm³ (20 °C)
MDL Number: MFCD00051520
CAS Number: 2905-23-9
EINECS: 220-807-3
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR