2,5-Dimethylthiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

002791-25G 002791-100G
FLUO002791-25GEA 0 GBP
FLUO002791-25G FLUO002791-100G
2,5-Dimethylthiophene
2,5-Dimethylthiophene
Formula: C₆H₈S
Boiling Pt: 134 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.984 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005452
CAS Number: 638-02-8
EINECS: 211-313-9
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR