3,4,5-Trifluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

002074-1G 002074-5G 002074-10G 002074-25G
FLUO002074-1GEA 0 GBP
FLUO002074-1G FLUO002074-5G FLUO002074-10G FLUO002074-25G
3,4,5-Trifluorophenol
3,4,5-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042427
CAS Number: 99627-05-1
EINECS: 436-120-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR