4-(Dimethylamino)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

001704-100G 001704-500G 001704-1KG
FLUO001704-100GEA 0 GBP
FLUO001704-100G FLUO001704-500G FLUO001704-1KG
4-(Dimethylamino)pyridine
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 109…111 °C
Flash Pt: 124 °C
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1,I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR